Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016726.hg.1 |
TC08001293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016726.hg.1 |
ENST00000262215.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016726.hg.1 |
ENST00000518230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016726.hg.1 |
ENST00000520381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016726.hg.1 |
NM_006421.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000262215.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000518230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000520381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
NM_006421.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000262215.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000518230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
ENST00000520381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
NM_006421.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172732.hg.1 |
TC0800010703.hg.1 |
Clariom D TC |