Assay Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024.3 | NP_001015.1 | 1 - 2 | 72 | 79 | |
| GenBank mRNA | BM840443.1 | - | 1 - 2 | 72 | 79 | |
| CF124568.1 | - | 1 - 2 | 113 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000489.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001024.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
poly(A) RNA binding
protein N-terminus binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
poly(A) RNA binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20006885.hg.1 | ENST00000343986.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006885.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006885.hg.1 | TC20000489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007228.hg.1 | ENST00000490739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008820.hg.1 | ENST00000451001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008820.hg.1 | ENST00000451001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000317311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000337102.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000343986.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000370592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000450116.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | ENST00000492356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006888.hg.1 | TC20000489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | ENST00000317311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | ENST00000343986.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | ENST00000370592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | ENST00000492356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006880.hg.1 | TC20000489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007937.hg.1 | ENST00000478593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008007.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007937.hg.1 | ENST00000478593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008820.hg.1 | ENST00000451001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008820.hg.1 | ENST00000451001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155727.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155727.hg.1 | TC2000008007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155727.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007228.hg.1 | ENST00000490739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | NM_001024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | TC2000008007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000343986.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000370562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000370592.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000450116.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | ENST00000492356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155737.hg.1 | NM_001024.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 158904 | - | - |

RPS21
Genomic Map