Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
TC03000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
ENST00000310271.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
ENST00000394965.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
ENST00000441729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
ENST00000469224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
ENST00000484952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
NM_000688.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007050.hg.1 |
NM_199166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000310271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000394965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000469224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000484952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_000688.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_001304443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_001304444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_199166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000310271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000394965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000469224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
ENST00000484952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_000688.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_001304443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_001304444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
NM_199166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153636.hg.1 |
TC0300007466.hg.1 |
Clariom D TC |