Assay Details
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002676.2 | NP_002667.2 | 4 - 5 | 440 | 79 | |
XM_011530230.2 | XP_011528532.1 | 5 - 6 | 497 | 79 | ||
GenBank mRNA | AK289368.1 | - | 4 - 5 | 413 | 79 | |
AK316580.1 | - | 4 - 5 | 382 | 79 | ||
BC010855.2 | - | 4 - 5 | 393 | 3898208 | 79 | |
BC016818.1 | - | 4 - 5 | 444 | 4096204 | 79 | |
CR456544.1 | - | 4 - 5 | 392 | 79 | ||
D87810.1 | - | 4 - 5 | 393 | 79 | ||
U62526.1 | - | 4 - 5 | 382 | 79 | ||
U86070.1 | - | 4 - 5 | 387 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000828.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22014024.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014024.hg.1 | ENST00000466645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014024.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014024.hg.1 | TC22000828.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | TC2200008831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200163984.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
