Assay Details
| Gene Symbol: CD47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001777.3 | NP_001768.1 | 6 - 7 | 961 | 106 | |
| NM_198793.2 | NP_942088.1 | 6 - 7 | 961 | 106 | ||
| XM_005247908.1 | XP_005247965.1 | 6 - 7 | 931 | 106 | ||
| XM_005247909.1 | XP_005247966.1 | 6 - 7 | 931 | 106 | ||
| GenBank mRNA | AI701980.1 | - | 4 - 5 | 106 | ||
| AK289813.1 | - | 6 - 7 | 902 | 106 | ||
| BC010016.2 | - | 6 - 7 | 863 | 4303534 | 106 | |
| BC012884.1 | - | 6 - 7 | 884 | 3857911 | 106 | |
| BC037306.1 | - | 6 - 7 | 898 | 5260986 | 106 | |
| BT006907.1 | - | 6 - 7 | 781 | 106 | ||
| LN680437.1 | - | 6 - 7 | 781 | 106 | ||
| X69398.1 | - | 6 - 7 | 887 | 106 | ||
| Z25521.1 | - | 6 - 7 | 920 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001629.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
integrin-mediated signaling pathway
opsonization
positive regulation of cell proliferation
response to bacterium
positive regulation of cell-cell adhesion
extracellular matrix organization
positive regulation of inflammatory response
positive regulation of phagocytosis
positive regulation of T cell activation
leukocyte migration
protein binding
thrombospondin receptor activity
cell adhesion
integrin-mediated signaling pathway
opsonization
positive regulation of cell proliferation
response to bacterium
positive regulation of cell-cell adhesion
extracellular matrix organization
positive regulation of inflammatory response
positive regulation of phagocytosis
positive regulation of T cell activation
leukocyte migration
protein binding
thrombospondin receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03029390.hg.1 | TC03001629.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029390.hg.1 | ENST00000355354.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029390.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029390.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029390.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188802.hg.1 | TC0300011936.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188803.hg.1 | TC0300011936.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |

CD47
Genomic Map