Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011513114.2 | XP_011511416.1 | 2 - 3 | 144 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011513114.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
