Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003262.3 | NP_003253.1 | 6 - 7 | 641 | 130 | |
XM_011513114.2 | XP_011511416.1 | 7 - 8 | 718 | 130 | ||
GenBank mRNA | BC012035.1 | - | 6 - 7 | 625 | 4706586 | 130 |
BX648539.1 | - | 6 - 7 | 625 | 130 | ||
D87127.1 | - | 6 - 7 | 622 | 130 | ||
U93239.1 | - | 6 - 7 | 622 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000909.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03015897.hg.1 | TC03000909.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015897.hg.1 | ENST00000337002.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015897.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015897.hg.1 | ENST00000480708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015897.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167940.hg.1 | TC0300009458.hg.1 | Clariom D TC |
