Assay Details
| Gene Symbol: SI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001041.3 | NP_001032.2 | 23 - 24 | 2620 | 137 | |
| XM_011513078.2 | XP_011511380.1 | 23 - 24 | 2624 | 137 | ||
| GenBank mRNA | BC115034.1 | - | 23 - 24 | 2633 | 7962025 | 137 |
| BC116452.1 | - | 23 - 24 | 2633 | 7962021 | 137 | |
| BC116453.1 | - | 23 - 24 | 2633 | 137 | ||
| BC132834.1 | - | 23 - 24 | 2613 | 40146856 | 137 | |
| BC132860.1 | - | 23 - 24 | 2613 | 40146882 | 137 | |
| X63597.1 | - | 23 - 24 | 2619 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

SI
Genomic Map