Assay Details
| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031725.5 | NP_001026895.2 | 1 - 2 | 233 | 86 | |
| NM_001320181.1 | NP_001307110.1 | 1 - 2 | 233 | 86 | ||
| XM_005245519.1 | XP_005245576.1 | 1 - 2 | 142 | 86 | ||
| XM_005245521.2 | XP_005245578.1 | 1 - 2 | 142 | 86 | ||
| XM_006711562.1 | XP_006711625.1 | 1 - 2 | 142 | 86 | ||
| GenBank mRNA | AK299219.1 | - | 1 - 2 | 104 | 86 | |
| AK307569.1 | - | 161 | 86 | |||
| AL136611.1 | - | 1 - 2 | 146 | 86 | ||
| BC014183.2 | - | 1 - 2 | 166 | 86 | ||
| BC041801.1 | - | 1 - 2 | 224 | 5302098 | 86 | |
| DB483119.1 | - | 1 - 2 | 170 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003680.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK299219.1,AL136611.1,BC014183.2,BC041801.1,NM_001031725.5,NM_001320181.1,XM_005245519.1,XM_005245521.2,XM_006711562.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | TC01003680.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | ENST00000331314.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | ENST00000367346.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | ENST00000367348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | ENST00000447706.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056376.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | TC01003680.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | ENST00000331314.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | ENST00000367346.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | ENST00000367348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | ENST00000447706.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056378.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056383.hg.1 | TC01003680.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056383.hg.1 | ENST00000367346.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056383.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056379.hg.1 | TC01003680.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056379.hg.1 | ENST00000331314.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056379.hg.1 | ENST00000367346.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056379.hg.1 | ENST00000447706.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056379.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056382.hg.1 | TC01003680.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056382.hg.1 | ENST00000331314.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056382.hg.1 | ENST00000367346.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056382.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227764.hg.1 | TC0100016893.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100227768.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227768.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227768.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227768.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227768.hg.1 | TC0100016893.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100227769.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227769.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227769.hg.1 | TC0100016893.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227767.hg.1 | TC0100016893.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |

DDX59
Genomic Map