Assay Details
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031725.5 | NP_001026895.2 | 5 - 6 | 1553 | 134 | |
NM_001320181.1 | NP_001307110.1 | 5 - 6 | 1553 | 134 | ||
XM_005245519.1 | XP_005245576.1 | 5 - 6 | 1462 | 134 | ||
XM_006711562.1 | XP_006711625.1 | 5 - 6 | 1462 | 134 | ||
XM_011510035.2 | XP_011508337.1 | 5 - 6 | 1486 | 134 | ||
XM_017002431.1 | XP_016857920.1 | 5 - 6 | 2241 | 134 | ||
XM_017002432.1 | XP_016857921.1 | 5 - 6 | 1726 | 134 | ||
GenBank mRNA | AK299219.1 | - | 5 - 6 | 1424 | 134 | |
AK307569.1 | - | 1481 | 134 | |||
AL136611.1 | - | 5 - 6 | 1466 | 134 | ||
BC041801.1 | - | 5 - 6 | 1544 | 5302098 | 134 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003680.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | TC01003680.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | ENST00000331314.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | ENST00000367346.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | ENST00000367348.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | ENST00000447706.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056363.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | TC01003680.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | ENST00000331314.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | ENST00000367346.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | ENST00000367348.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | ENST00000447706.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01056364.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227750.hg.1 | TC0100016893.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000331314.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000413408.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000429498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000433235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | ENST00000447706.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | NM_001031725.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100227751.hg.1 | TC0100016893.hg.1 | Clariom D TC |
