Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
TC02000928.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
ENST00000280115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
ENST00000409029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
ENST00000409642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
ENST00000409876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
ENST00000498249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013594.hg.1 |
NM_177964.4 |
Refseq |