Assay Details
| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005711.4 | NP_005702.3 | 3 - 4 | 716 | 102 | |
| GenBank mRNA | AK312858.1 | - | 3 - 4 | 233 | 102 | |
| BC030828.1 | - | 3 - 4 | 642 | 4791845 | 102 | |
| U70312.1 | - | 3 - 4 | 333 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001558.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05021271.hg.1 | TC05001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021271.hg.1 | ENST00000296591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021271.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021271.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178609.hg.1 | TC0500011352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000296591.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000380138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000507663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | ENST00000510271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_001278642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011352.hg.1 | NM_005711.4 | Refseq |

EDIL3
Genomic Map