Assay Details
| Gene Symbol: RGS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081955.2 | NP_001075424.1 | 8 - 9 | 782 | 151 | |
| NM_001165933.1 | NP_001159405.1 | 8 - 9 | 782 | 151 | ||
| GenBank mRNA | AK290535.1 | - | 8 - 9 | 651 | 151 | |
| AK309061.1 | - | 8 - 9 | 672 | 151 | ||
| AY585191.1 | - | 8 - 9 | 579 | 151 | ||
| BC022504.1 | - | 8 - 9 | 649 | 4801328 | 151 | |
More Information
Set Membership: |
|

RGS9
Genomic Map