Assay Details
| Gene Symbol: ACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004300.3 | NP_004291.1 | 4 - 5 | 384 | 142 | |
| GenBank mRNA | AB209838.1 | - | 4 - 5 | 309 | 142 | |
| AK291861.1 | - | 4 - 5 | 312 | 142 | ||
| BC106011.1 | - | 4 - 5 | 290 | 4995529 | 142 | |
| BI460080.1 | - | 4 - 5 | 382 | 142 | ||
| DB443811.1 | - | 4 - 5 | 363 | 142 | ||
| M83653.1 | - | 4 - 5 | 332 | 142 | ||
| M87545.1 | - | 3 - 4 | 259 | 142 | ||
| S62884.1 | - | 3 - 4 | 259 | 142 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ACP1
Genomic Map