| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003770.hg.1 |
TC20001730.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003770.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
TC2000009915.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169736.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000342422.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000344795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000373852.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000483815.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000491113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000492721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
ENST00000494506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_001199534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_018840.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NM_199483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009915.hg.1 |
NR_026562.3 |
Refseq |