Assay Details
Gene Symbol: MRPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KU178614.1 | - | 1 - 2 | 192 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000276585.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000518371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000519386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000520946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000521605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | ENST00000522987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012432.hg.1 | NM_014018.2 | Refseq |
