Assay Details
Gene Symbol: KIAA1804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032435.2 | NP_115811.2 | 2 - 3 | 1245 | 141 | |
XM_011544305.2 | XP_011542607.1 | 2 - 3 | 1281 | 141 | ||
GenBank mRNA | BC136648.1 | - | 2 - 3 | 1104 | 9020638 | 141 |
BC136649.1 | - | 2 - 3 | 1104 | 9020639 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001927.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01029940.hg.1 | TC01001927.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01029940.hg.1 | ENST00000366623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029940.hg.1 | ENST00000366624.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029940.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01029941.hg.1 | TC01001927.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01029941.hg.1 | ENST00000366623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029941.hg.1 | ENST00000366624.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029941.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01029938.hg.1 | TC01001927.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01029938.hg.1 | ENST00000366623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029938.hg.1 | ENST00000366624.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029938.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01029939.hg.1 | TC01001927.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01029939.hg.1 | ENST00000366623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029939.hg.1 | ENST00000366624.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029939.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188535.hg.1 | TC0100012002.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188540.hg.1 | TC0100012002.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188536.hg.1 | TC0100012002.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188538.hg.1 | TC0100012002.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012002.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | ENST00000366623.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | ENST00000366624.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | NM_032435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100188534.hg.1 | TC0100012002.hg.1 | Clariom D TC |
