Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029944.hg.1 |
TC01001927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029944.hg.1 |
ENST00000366623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029944.hg.1 |
ENST00000366624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029944.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012002.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
ENST00000366623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
ENST00000366624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
NM_032435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188545.hg.1 |
TC0100012002.hg.1 |
Clariom D TC |