Assay Details
Gene Symbol: CEP78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098802.1 | NP_001092272.1 | 15 - 16 | 2127 | 74 | |
XM_005252263.4 | XP_005252320.1 | 15 - 16 | 2140 | 74 | ||
XM_005252266.4 | XP_005252323.1 | 15 - 16 | 2134 | 74 | ||
XM_017015192.1 | XP_016870681.1 | 15 - 16 | 2137 | 74 | ||
XM_017015193.1 | XP_016870682.1 | 15 - 16 | 2137 | 74 | ||
XM_017015196.1 | XP_016870685.1 | 15 - 16 | 2131 | 74 | ||
GenBank mRNA | BC128058.1 | - | 15 - 16 | 1995 | 40078333 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
