Assay Details
| Gene Symbol: SLC20A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005415.4 | NP_005406.3 | 8 - 9 | 2143 | 58 | |
| XM_017004768.1 | XP_016860257.1 | 8 - 9 | 1892 | 58 | ||
| XM_017004769.1 | XP_016860258.1 | 9 - 10 | 4370 | 58 | ||
| XM_017004770.1 | XP_016860259.1 | 9 - 10 | 2173 | 58 | ||
| XM_017004771.1 | XP_016860260.1 | 10 - 11 | 2209 | 58 | ||
| GenBank mRNA | AK123420.1 | - | 3 - 4 | 1681 | 58 | |
| AK312738.1 | - | 8 - 9 | 2062 | 58 | ||
| BC019944.1 | - | 8 - 9 | 2050 | 3918690 | 58 | |
| BC075818.1 | - | 2 - 3 | 652 | 58 | ||
| BX647555.1 | - | 1 - 2 | 1396 | 58 | ||
| L20859.1 | - | 8 - 9 | 1974 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000711.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphate-containing compound metabolic process
transport
ion transport
signal transduction
phosphate ion transmembrane transport
sodium ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
sodium-dependent phosphate transport
signal transducer activity
receptor activity
inorganic phosphate transmembrane transporter activity
high-affinity inorganic phosphate:sodium symporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
phosphate-containing compound metabolic process
transport
ion transport
signal transduction
phosphate ion transmembrane transport
sodium ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
sodium-dependent phosphate transport
signal transducer activity
receptor activity
inorganic phosphate transmembrane transporter activity
high-affinity inorganic phosphate:sodium symporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02011046.hg.1 | TC02000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011046.hg.1 | ENST00000272542.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011046.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011044.hg.1 | TC02000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011044.hg.1 | ENST00000272542.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011044.hg.1 | ENST00000409095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011044.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | ENST00000272542.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | ENST00000490674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | NM_005415.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162389.hg.1 | TC0200008930.hg.1 | Clariom D TC |

SLC20A1
Genomic Map