Assay Details
| Gene Symbol: AGPAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248805.3 | XP_005248862.1 | 1 - 2 | 632 | 84 | |
| GenBank mRNA | AK056346.1 | - | 1 - 2 | 836 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056346.1, XM_005248805.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
positive regulation of cellular metabolic process
1-acylglycerol-3-phosphate O-acyltransferase activity
protein binding
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
positive regulation of cellular metabolic process
1-acylglycerol-3-phosphate O-acyltransferase activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000336984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375104.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000375107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000395499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000476663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000490711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | ENST00000617181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_006411.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011481.hg.1 | NM_032741.4 | Refseq |

AGPAT1
Genomic Map