Assay Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018046.4 | NP_060516.2 | 4 - 5 | 1053 | 155 | |
| GenBank mRNA | AW958325.1 | - | 4 - 5 | 535 | 155 | |
| AY500994.1 | - | 4 - 5 | 972 | 155 | ||
| BC032844.1 | - | 4 - 5 | 1039 | 5266978 | 155 | |
| BC073864.1 | - | 4 - 5 | 580 | 155 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000377.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05005274.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005274.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005274.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005273.hg.1 | TC05000377.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005273.hg.1 | ENST00000312916.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005273.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153669.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153670.hg.1 | TC0500007841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |

AGGF1
Genomic Map