Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC012035.1 | - | 8 - 9 | 1254 | 4706586 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167943.hg.1 | TC0300009458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167945.hg.1 | TC0300009458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
