| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
TC03000620.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
ENST00000273666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
ENST00000471454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
ENST00000472879.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
ENST00000492541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
ENST00000497029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011221.hg.1 |
NM_014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000273666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000471262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000471454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000472879.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000492541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000497029.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
NM_001308330.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
NM_014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000273666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000471262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000471454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000472879.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000492541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
ENST00000497029.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
NM_001308330.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
NM_014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160675.hg.1 |
TC0300008524.hg.1 |
Clariom D TC |