Assay Details
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282801.1 | NP_001269730.1 | 5 - 6 | 595 | 120 | |
NM_001282802.1 | NP_001269731.1 | 5 - 6 | 743 | 120 | ||
NM_020654.4 | NP_065705.3 | 6 - 7 | 793 | 120 | ||
XM_011513038.2 | XP_011511340.1 | 6 - 7 | 782 | 120 | ||
XM_011513039.2 | XP_011511341.1 | 5 - 6 | 707 | 120 | ||
XM_011513041.2 | XP_011511343.1 | 5 - 6 | 584 | 120 | ||
XM_011513045.2 | XP_011511347.1 | 6 - 7 | 783 | 120 | ||
XM_017006924.1 | XP_016862413.1 | 6 - 7 | 670 | 120 | ||
XM_017006925.1 | XP_016862414.1 | 6 - 7 | 758 | 120 | ||
XM_017006927.1 | XP_016862416.1 | 4 - 5 | 493 | 120 | ||
XM_017006928.1 | XP_016862417.1 | 6 - 7 | 783 | 120 | ||
GenBank mRNA | AA651903.1 | - | 1 - 2 | 120 | ||
AF199458.1 | - | 5 - 6 | 741 | 120 | ||
AK297356.1 | - | 704 | 120 | |||
AL136599.1 | - | 5 - 6 | 621 | 120 | ||
BC062674.1 | - | 6 - 7 | 740 | 120 | ||
BC129988.1 | - | 6 - 7 | 720 | 40128878 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
