Assay Details
| Gene Symbol: P3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271110.2 | XP_005271167.1 | 1 - 2 | 69 | 89 | |
| XM_011541948.1 | XP_011540250.1 | 1 - 2 | 69 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005271110.2, XM_011541948.1 transcripts.
Set Membership: |
|

P3H1
Genomic Map