Assay Details
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093418.1 | - | 11 - 12 | 1197 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000255858.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000320982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000321205.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | ENST00000381982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_001161368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008444.hg.1 | NM_174977.3 | Refseq |
