Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000808.3 | NP_000799.1 | 2 - 3 | 383 | 121 | |
| XM_006724811.2 | XP_006724874.1 | 2 - 3 | 383 | 121 | ||
| GenBank mRNA | A28102.1 | - | 2 - 3 | 229 | 121 | |
| AK312404.1 | - | 2 - 3 | 361 | 121 | ||
| BC028629.1 | - | 2 - 3 | 382 | 5261326 | 121 | |
| DA402645.1 | - | 2 - 3 | 383 | 121 | ||
| KJ620007.1 | - | 8 - 9 | 1012 | 121 | ||
| S62908.1 | - | 2 - 3 | 228 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001493.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
benzodiazepine receptor activity
transport
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
benzodiazepine receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000417858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000417858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000497894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000497894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019472.hg.1 | TC0X001493.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019472.hg.1 | ENST00000370311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019472.hg.1 | ENST00000370314.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019472.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019472.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00175414.hg.1 | TC0X00011126.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000370314.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000417858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000417858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000497894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000497894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | ENST00000535043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011126.hg.1 | NM_000808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |

GABRA3
Genomic Map