| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005484.hg.1 |
TC01000353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005484.hg.1 |
ENST00000339276.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005484.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005487.hg.1 |
TC01000353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005487.hg.1 |
ENST00000339276.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005487.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005489.hg.1 |
TC01000353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005489.hg.1 |
ENST00000339276.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005489.hg.1 |
ENST00000538651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005489.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005485.hg.1 |
TC01000353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005485.hg.1 |
ENST00000339276.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005485.hg.1 |
ENST00000538651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005485.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005490.hg.1 |
TC01000353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005490.hg.1 |
ENST00000339276.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005490.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152388.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152388.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152388.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152388.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152388.hg.1 |
TC0100007486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100152389.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152389.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152389.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152389.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152389.hg.1 |
TC0100007486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007486.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152390.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152390.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152390.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152390.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152390.hg.1 |
TC0100007486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100152391.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152391.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152391.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152391.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152391.hg.1 |
TC0100007486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100152392.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152392.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152392.hg.1 |
ENST00000339276.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152392.hg.1 |
NM_006142.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152392.hg.1 |
TC0100007486.hg.1 |
Clariom D TC |