Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
TC07000633.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000348028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000349350.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000421100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000427578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000443370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000449052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000538115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000542585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000546213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
ENST00000546292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
NM_003386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009724.hg.1 |
NM_173059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000538115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000542585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000546213.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000546292.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000613979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000618565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000620596.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000620868.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NM_003386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NM_173059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111917.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000538115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000542585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000546213.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000546292.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000613979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000618565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000620596.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
ENST00000620868.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NM_003386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NM_173059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111917.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
NR_111919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160406.hg.1 |
TC0700008564.hg.1 |
Clariom D TC |