Assay Details
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005443.4 | NP_005434.4 | 4 - 5 | 633 | 98 | |
XM_011532400.1 | XP_011530702.1 | 4 - 5 | 613 | 98 | ||
XM_011532401.1 | XP_011530703.1 | 4 - 5 | 699 | 98 | ||
GenBank mRNA | AF016496.1 | - | 4 - 5 | 549 | 98 | |
AF033026.1 | - | 4 - 5 | 575 | 98 | ||
AF105227.1 | - | 4 - 5 | 603 | 98 | ||
AK292774.1 | - | 4 - 5 | 583 | 98 | ||
AU132232.1 | - | 4 - 5 | 633 | 98 | ||
BC011392.1 | - | 4 - 5 | 506 | 3869484 | 98 | |
BC032513.1 | - | 4 - 5 | 602 | 98 | ||
BC040940.1 | - | 4 - 5 | 561 | 98 | ||
BC050627.1 | - | 4 - 5 | 569 | 5556165 | 98 | |
BQ437924.1 | - | 4 - 5 | 561 | 98 | ||
CR457028.1 | - | 4 - 5 | 549 | 98 | ||
Y10387.1 | - | 4 - 5 | 585 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001455.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04020555.hg.1 | TC04001455.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04020555.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020555.hg.1 | ENST00000511304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020555.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | ENST00000514815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04020551.hg.1 | TC04001455.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04020551.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020551.hg.1 | ENST00000511304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020551.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04020553.hg.1 | TC04001455.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04020553.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020553.hg.1 | ENST00000511304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04020553.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179204.hg.1 | TC0400011538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179200.hg.1 | TC0400011538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000504987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000506544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000512641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | ENST00000514815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011538.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000265174.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000502431.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | ENST00000511304.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | NM_005443.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400179202.hg.1 | TC0400011538.hg.1 | Clariom D TC |
