Assay Details
| Gene Symbol: MRPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021107.1 | NP_066930.1 | 1 - 2 | 391 | 80 | |
| NM_033362.3 | NP_203526.1 | 2 - 3 | 360 | 80 | ||
| NM_033363.1 | NP_203527.1 | 2 - 3 | 287 | 80 | ||
| GenBank mRNA | AI168197.1 | - | 1 - 2 | 80 | ||
| BC001617.1 | - | 2 - 3 | 242 | 3547013 | 80 | |
| BC006424.1 | - | 2 - 3 | 520 | 3627808 | 80 | |
| BC007499.2 | - | 2 - 3 | 113 | 2959394 | 80 | |
| CR407609.1 | - | 1 - 2 | 50 | 80 | ||
| Y11681.1 | - | 1 - 2 | 391 | 80 | ||
More Information
Set Membership: |
|

MRPS12
Genomic Map