Assay Details
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278636.1 | NP_001265565.1 | 1 - 2 | 237 | 76 | |
NM_032565.4 | NP_115954.1 | 1 - 2 | 237 | 76 | ||
GenBank mRNA | AF243433.1 | - | 1 - 2 | 226 | 76 | |
BC018478.1 | - | 1 - 2 | 203 | 4476654 | 76 | |
BC021021.1 | - | 1 - 2 | 214 | 76 | ||
BC092471.1 | - | 1 - 2 | 232 | 4796715 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000662.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000242827.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000378272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000378274.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000378282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000378284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | ENST00000495963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007645.hg.1 | TC13000662.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000242827.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378274.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000378284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | ENST00000495963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007648.hg.1 | TC13000662.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000242827.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378274.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000378284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | ENST00000495963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007647.hg.1 | TC13000662.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160243.hg.1 | TC1300008922.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008922.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010557.hg.1 | ENST00000569153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000242827.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378270.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378272.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000378284.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000473576.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | ENST00000495963.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NM_001278636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NM_032565.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NR_103802.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | NR_103803.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300160245.hg.1 | TC1300008922.hg.1 | Clariom D TC |
