| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015111.hg.1 |
ENST00000330817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015111.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015111.hg.1 |
TC22000904.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000486602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
ENST00000492791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009128.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165968.hg.1 |
TC2200009128.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165968.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165968.hg.1 |
NM_024105.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165968.hg.1 |
ENST00000330817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165968.hg.1 |
NM_024105.3 |
Refseq |