Assay Details
Gene Symbol: GALNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017423.2 | NP_059119.2 | 8 - 9 | 1468 | 78 | |
XM_005263062.1 | XP_005263119.1 | 8 - 9 | 1468 | 78 | ||
XM_011532025.2 | XP_011530327.1 | 8 - 9 | 2215 | 78 | ||
XM_017008292.1 | XP_016863781.1 | 11 - 12 | 2388 | 78 | ||
XM_017008293.1 | XP_016863782.1 | 11 - 12 | 4177 | 78 | ||
GenBank mRNA | AJ002744.1 | - | 8 - 9 | 1417 | 78 | |
AK295509.1 | - | 6 - 7 | 774 | 78 | ||
BC035303.1 | - | 7 - 8 | 899 | 78 | ||
BC046129.1 | - | 8 - 9 | 1446 | 4442080 | 78 | |
BC047468.1 | - | 8 - 9 | 1443 | 5271749 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000853.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04011729.hg.1 | TC04000853.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04011729.hg.1 | ENST00000265000.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011729.hg.1 | ENST00000458613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011729.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04011728.hg.1 | TC04000853.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04011728.hg.1 | ENST00000265000.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011728.hg.1 | ENST00000458613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011728.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163957.hg.1 | TC0400009322.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163958.hg.1 | TC0400009322.hg.1 | Clariom D TC |
