Assay Details
Gene Symbol: GALNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017423.2 | NP_059119.2 | 9 - 10 | 1694 | 87 | |
XM_005263062.1 | XP_005263119.1 | 9 - 10 | 1694 | 87 | ||
XM_011532025.2 | XP_011530327.1 | 9 - 10 | 2441 | 87 | ||
XM_017008292.1 | XP_016863781.1 | 12 - 13 | 2614 | 87 | ||
XM_017008293.1 | XP_016863782.1 | 12 - 13 | 4403 | 87 | ||
GenBank mRNA | AJ002744.1 | - | 9 - 10 | 1643 | 87 | |
AJ505958.1 | - | 1 - 2 | 138 | 87 | ||
AK295509.1 | - | 7 - 8 | 1000 | 87 | ||
BC035303.1 | - | 8 - 9 | 1125 | 87 | ||
BC046129.1 | - | 9 - 10 | 1672 | 4442080 | 87 | |
BC047468.1 | - | 9 - 10 | 1669 | 5271749 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000853.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04011731.hg.1 | TC04000853.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04011731.hg.1 | ENST00000265000.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011731.hg.1 | ENST00000458613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011731.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000265000.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000503213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000505308.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | ENST00000515862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | NM_017423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163960.hg.1 | TC0400009322.hg.1 | Clariom D TC |
