Assay Details
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317841.1 | NP_001304770.1 | 1 - 2 | 372 | 79 | |
NM_152399.3 | NP_689612.3 | 1 - 2 | 372 | 79 | ||
XM_017007725.1 | XP_016863214.1 | 1 - 2 | 89 | 79 | ||
XM_017007726.1 | XP_016863215.1 | 1 - 2 | 89 | 79 | ||
GenBank mRNA | BC047657.1 | - | 1 - 2 | 372 | 5264721 | 79 |
HY117228.1 | - | 1 - 2 | 465 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001515.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC047657.1,NM_001317841.1,NM_152399.3,XM_017007725.1,XM_017007726.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04021526.hg.1 | TC04001515.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021526.hg.1 | ENST00000337677.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021526.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021525.hg.1 | TC04001515.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021525.hg.1 | ENST00000337677.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021525.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021519.hg.1 | TC04001515.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021519.hg.1 | ENST00000337677.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021519.hg.1 | ENST00000394394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021519.hg.1 | ENST00000394396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021519.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180809.hg.1 | TC0400011746.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400180810.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180810.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180810.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180810.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180810.hg.1 | TC0400011746.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180804.hg.1 | TC0400011746.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 113407 | - | - |
