Assay Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF070642.1 | - | 4 - 5 | 767 | 67 | |
| BC126321.1 | - | 4 - 5 | 1394 | 8992037 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000308748.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000483405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000563177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007431.hg.1 | NM_181718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153883.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153883.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153883.hg.1 | ENST00000414952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153883.hg.1 | ENST00000566693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153883.hg.1 | TC1600007431.hg.1 | Clariom D TC |

ASPHD1
Genomic Map