Assay Details
| Gene Symbol: GUCY1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003923.2 | 12 - 13 | 1877 | 107 | ||
| GenBank mRNA | AF038499.2 | - | 12 - 13 | 1726 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease H2 subunit B | RNASEH2B |
AGS2, DLEU8 |
-39.6kb | Anti-sense | Chr. 13 - 50909678 - 50970462 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 371 | LINC00371 | LINC00372 | 66.5kb | Sense | Chr. 13 - 51082848 - 51172388 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000675.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007787.hg.1 | ENST00000493639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007787.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007787.hg.1 | TC13000675.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160543.hg.1 | TC1300008971.hg.1 | Clariom D TC |

GUCY1B2
Genomic Map