Assay Details
| Gene Symbol: GUCY1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003923.2 | 16 - 17 | 126 | |||
| GenBank mRNA | AF038499.2 | - | 16 - 17 | 126 | ||
| AI247180.1 | - | 1 - 2 | 306 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000675.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13007780.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007780.hg.1 | TC13000675.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007781.hg.1 | ENST00000493639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007781.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007781.hg.1 | TC13000675.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007782.hg.1 | ENST00000493639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007782.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007782.hg.1 | TC13000675.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160534.hg.1 | TC1300008971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300160532.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160532.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160532.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160532.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160532.hg.1 | TC1300008971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160533.hg.1 | TC1300008971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198091 | - | - |

GUCY1B2
Genomic Map