Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000308475.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000372702.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000394707.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000479884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000491677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000494588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000607131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000607487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
NM_001007271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
NM_001007273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000308475.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000372702.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000394707.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000479884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000491677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000494588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000607131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
ENST00000607487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
NM_001007271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
NM_001007273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175522.hg.1 |
TC1000011072.hg.1 |
Clariom D TC |