Assay Details
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135120.1 | 5 - 6 | 712 | 152 | ||
NR_135121.1 | 5 - 6 | 610 | 152 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
