Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014125.hg.1 |
TC09001059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014125.hg.1 |
ENST00000314888.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014125.hg.1 |
ENST00000540444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014125.hg.1 |
NM_006289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168497.hg.1 |
ENST00000314888.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168497.hg.1 |
NM_006289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168497.hg.1 |
ENST00000314888.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168497.hg.1 |
NM_006289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168497.hg.1 |
TC0900009950.hg.1 |
Clariom D TC |