Assay Details
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006981.3 | NP_008912.2 | 5 - 6 | 1987 | 102 | |
NM_173200.2 | NP_775292.1 | 6 - 7 | 2059 | 102 | ||
XM_017015162.1 | XP_016870651.1 | 4 - 5 | 1919 | 102 | ||
GenBank mRNA | AK226108.1 | - | 5 - 6 | 1969 | 102 | |
D78579.1 | - | 5 - 6 | 1989 | 102 | ||
S81243.1 | - | 5 - 6 | 1874 | 102 | ||
U12767.1 | - | 4 - 5 | 1350 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000508.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mesoderm formation
transcription initiation from RNA polymerase II promoter
gastrulation
axon guidance
biological_process
pyruvate oxidation
positive regulation of cardiac muscle hypertrophy
positive regulation of glucose transport
hippocampus development
intracellular receptor signaling pathway
adult behavior
organ regeneration
positive regulation of mast cell cytokine production
common myeloid progenitor cell proliferation
positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
response to hydrogen peroxide
mast cell degranulation
steroid hormone mediated signaling pathway
negative regulation of neuron apoptotic process
cellular response to leptin stimulus
cellular respiration
fat cell differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
regulation of smooth muscle cell proliferation
positive regulation of smooth muscle cell proliferation
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
neuromuscular process controlling balance
vestibular reflex
regulation of type B pancreatic cell proliferation
cellular response to corticotropin-releasing hormone stimulus
cellular response to catecholamine stimulus
energy homeostasis
positive regulation of monocyte aggregation
negative regulation of hydrogen peroxide-induced neuron death
positive regulation of leukocyte apoptotic process
positive regulation of feeding behavior
regulation of energy homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
protein kinase binding
histone acetyltransferase binding
glucocorticoid receptor binding
protein homodimerization activity
negative regulation of transcription from RNA polymerase II promoter
mesoderm formation
transcription initiation from RNA polymerase II promoter
gastrulation
axon guidance
biological_process
pyruvate oxidation
positive regulation of cardiac muscle hypertrophy
positive regulation of glucose transport
hippocampus development
intracellular receptor signaling pathway
adult behavior
organ regeneration
positive regulation of mast cell cytokine production
common myeloid progenitor cell proliferation
positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
response to hydrogen peroxide
mast cell degranulation
steroid hormone mediated signaling pathway
negative regulation of neuron apoptotic process
cellular response to leptin stimulus
cellular respiration
fat cell differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
regulation of smooth muscle cell proliferation
positive regulation of smooth muscle cell proliferation
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
neuromuscular process controlling balance
vestibular reflex
regulation of type B pancreatic cell proliferation
cellular response to corticotropin-releasing hormone stimulus
cellular response to catecholamine stimulus
energy homeostasis
positive regulation of monocyte aggregation
negative regulation of hydrogen peroxide-induced neuron death
positive regulation of leukocyte apoptotic process
positive regulation of feeding behavior
regulation of energy homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
protein kinase binding
histone acetyltransferase binding
glucocorticoid receptor binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09005520.hg.1 | TC09000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09005520.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09005520.hg.1 | ENST00000395097.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09005520.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09005520.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000338488.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173199.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008219.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000330847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000395097.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | ENST00000618101.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | NM_006981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | NM_173200.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900155063.hg.1 | TC0900008219.hg.1 | Clariom D TC |
