Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
TC04001815.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
ENST00000328559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
ENST00000362004.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
ENST00000505916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
ENST00000510776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024757.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191509.hg.1 |
TC0400013001.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000328559.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000510776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
ENST00000511404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400013001.hg.1 |
NM_018409.3 |
Refseq |