Assay Details
| Gene Symbol: ARG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC005321.1 | - | 5 - 6 | 611 | 3934479 | 140 |
| BT006741.1 | - | 5 - 6 | 554 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164846.hg.1 | TC0600009455.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164847.hg.1 | TC0600009455.hg.1 | Clariom D TC |

ARG1
Genomic Map