Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011634.hg.1 |
TC09000874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011634.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011634.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164603.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900164603.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164603.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900164603.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164603.hg.1 |
TC0900009404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000382032.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000397885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000469168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
ENST00000490444.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009404.hg.1 |
NM_014878.4 |
Refseq |