Assay Details
Gene Symbol: SELO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031454.1 | NP_113642.1 | 4 - 5 | 1130 | 68 | |
GenBank mRNA | AY324823.1 | - | 4 - 5 | 1130 | 68 | |
BC001099.2 | - | 4 - 5 | 620 | 68 | ||
BC020510.1 | - | 3 - 4 | 441 | 68 | ||
BC110866.1 | - | 4 - 5 | 1100 | 6041782 | 68 | |
CR456464.1 | - | 4 - 5 | 646 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000421.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22008011.hg.1 | ENST00000380903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22008011.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22008011.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22008011.hg.1 | TC22000421.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | TC2200007792.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200155338.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000380903.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000492092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | ENST00000611222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007792.hg.1 | NM_031454.1 | Refseq |
