Assay Details
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033025.5 | NP_149014.3 | 1 - 2 | 156 | 159 | |
GenBank mRNA | AK123686.1 | - | 1 - 2 | 117 | 159 | |
HY178244.1 | - | 1 - 2 | 157 | 159 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000269.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19003834.hg.1 | ENST00000342784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19003834.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19003834.hg.1 | TC19000269.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19003831.hg.1 | ENST00000342784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19003831.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19003831.hg.1 | TC19000269.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19003833.hg.1 | ENST00000342784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19003833.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19003833.hg.1 | TC19000269.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152060.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152060.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152060.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152060.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152060.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152064.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152066.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152067.hg.1 | NM_033025.5 | Refseq |
