Assay Details
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300910.1 | NP_001287839.1 | 2 - 3 | 303 | 76 | |
NM_033025.5 | NP_149014.3 | 2 - 3 | 504 | 76 | ||
GenBank mRNA | AK123686.1 | - | 2 - 3 | 465 | 76 | |
AK315453.1 | - | 2 - 3 | 262 | 76 | ||
BC029926.1 | - | 2 - 3 | 245 | 5106728 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000269.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19003836.hg.1 | ENST00000342784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19003836.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19003836.hg.1 | TC19000269.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152072.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | NM_033025.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | TC1900007208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000342784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000597977.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000600252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | ENST00000600440.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | NM_001300910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900152069.hg.1 | NM_033025.5 | Refseq |
